Product Details

SNP ID
rs188023685
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32394763 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAACCTGAGAGAGAAAAAGTTGC[A/G]ATTTTCTCCTCGCCCAAAAAGGGGA
Phenotype
MIM: 606000
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BTNL2 PubMed Links

Gene Details

Gene
BTNL2
Gene Name
butyrophilin like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304561.1 3097 Silent Mutation ATC,ATT I447I NP_001291490.1
XM_017011057.1 3097 Silent Mutation ATC,ATT I447I XP_016866546.1
Gene
HCG23
Gene Name
HLA complex group 23 (non-protein coding)
There are no transcripts associated with this gene.

Gene
LOC101929163
Gene Name
uncharacterized LOC101929163
There are no transcripts associated with this gene.

View Full Product Details