Product Details

SNP ID
rs189789985
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66043101 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCGGCGATCATGACGAGCGAGAA[A/G]ACCTCCTCCACCTGGCGGATGAGGC
Phenotype
MIM: 608234 MIM: 605591
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GAL3ST3 PubMed Links

Gene Details

Gene
GAL3ST3
Gene Name
galactose-3-O-sulfotransferase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033036.2 898 Silent Mutation GTC,GTT V234V NP_149025.1
XM_017018519.1 898 Silent Mutation GTC,GTT V256V XP_016874008.1
Gene
SF3B2
Gene Name
splicing factor 3b subunit 2
There are no transcripts associated with this gene.

View Full Product Details