Product Details

SNP ID
rs190453542
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:231593110 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGCAGGATATAGATCAGGCCTC[C/T]TTCAAGACGGCCACCCCCAGGGCCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C2orf57 PubMed Links

Gene Details

Gene
C2orf57
Gene Name
chromosome 2 open reading frame 57
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152614.2 247 Silent Mutation TCC,TCT S53S NP_689827.2

View Full Product Details