Product Details

SNP ID
rs190177424
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:102651156 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAGAAGTGGCTGGTACCTGTGTG[C/G]GTCCTCGTGTGGGCCTTCAGATGGG
Phenotype
MIM: 601878
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KLF10 PubMed Links

Gene Details

Gene
KLF10
Gene Name
Kruppel like factor 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032282.3 1477 Silent Mutation ACC,ACG T381T NP_001027453.1
NM_005655.3 1477 Silent Mutation ACC,ACG T392T NP_005646.1
Gene
LOC101927245
Gene Name
vegetative cell wall protein gp1-like
There are no transcripts associated with this gene.

View Full Product Details