Product Details

SNP ID
rs191585865
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:27836889 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAATTCAGAAAACACAGAAGAGGA[A/T]AAAAAAGAACAGACACTATCACTGT
Phenotype
MIM: 606246 MIM: 605326
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
JAZF1 PubMed Links

Gene Details

Gene
JAZF1
Gene Name
JAZF zinc finger 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175061.3 Intron NP_778231.2
XM_006715656.1 Intron XP_006715719.1
XM_017011831.1 Intron XP_016867320.1
XM_017011832.1 Intron XP_016867321.1
Gene
TAX1BP1
Gene Name
Tax1 binding protein 1
There are no transcripts associated with this gene.

View Full Product Details