Product Details

SNP ID
rs191936506
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:72056190 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGCATGCTCTGCAAATCAAACCT[A/C]CAACATCTCCAAGTACAGGGGCAGA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C9orf57 PubMed Links

Gene Details

Gene
C9orf57
Gene Name
chromosome 9 open reading frame 57
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128618.1 170 Missense Mutation GGA,GTA G77V NP_001122090.1
XM_011518225.2 170 Missense Mutation GGA,GTA G27V XP_011516527.1

View Full Product Details