Product Details

SNP ID
rs192377800
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:102730904 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAAGTAACAAAATTCTGTTTTGT[C/T]GTTTTGGTCCTGTGAAACCAGGATG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC168 PubMed Links

Gene Details

Gene
CCDC168
Gene Name
coiled-coil domain containing 168
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146197.1 19933 Missense Mutation CAA,CGA Q6598R NP_001139669.1
XM_011521106.1 19933 Missense Mutation CAA,CGA Q6558R XP_011519408.1

View Full Product Details