Product Details

SNP ID
rs193198551
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:152085376 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTGACCAGGGGACTGAAGAATTG[C/T]GCTCTTTCTTGGCTGGACTTTTTTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100131107 PubMed Links

Gene Details

Gene
LOC100131107
Gene Name
putative UPF0607 protein ENSP00000383783
There are no transcripts associated with this gene.

Gene
TCHHL1
Gene Name
trichohyalin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008536.1 2371 Missense Mutation CAC,CGC H769R NP_001008536.1

View Full Product Details