Product Details

SNP ID
rs192734594
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:206909790 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGATGTTGTTTTGGGAGCGCGGT[C/G]GCGGCGACCCGCGGGGCCTCTGGGA
Phenotype
MIM: 606015 MIM: 604136
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FCMR PubMed Links

Gene Details

Gene
FCMR
Gene Name
Fc fragment of IgM receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142473.1 1099 Missense Mutation CCA,CGA P195R NP_001135945.1
NM_001193338.1 1099 Silent Mutation GCC,GCG A263A NP_001180267.1
NM_005449.4 1099 Missense Mutation CCA,CGA P307R NP_005440.1
XM_005273351.3 1099 Missense Mutation CCA,CGA P307R XP_005273408.1
XM_005273352.4 1099 Missense Mutation CCA,CGA P216R XP_005273409.1
Gene
IL24
Gene Name
interleukin 24
There are no transcripts associated with this gene.

View Full Product Details