Product Details

SNP ID
rs6043845
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:16272450 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGATTCTGTAACAAAAAATGTGAA[C/T]AATACTTAAAAATATGCATTTAGCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIF16B PubMed Links

Gene Details

Gene
KIF16B
Gene Name
kinesin family member 16B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199865.1 4788 UTR 3 NP_001186794.1
NM_001199866.1 4788 Intron NP_001186795.1
NM_024704.4 4788 UTR 3 NP_078980.3
XM_005260750.3 4788 Intron XP_005260807.1
XM_005260751.4 4788 Intron XP_005260808.1
XM_005260752.3 4788 Intron XP_005260809.1
XM_005260753.3 4788 UTR 3 XP_005260810.1
XM_005260754.3 4788 UTR 3 XP_005260811.1
XM_005260755.3 4788 UTR 3 XP_005260812.1
XM_006723588.3 4788 UTR 3 XP_006723651.1
XM_017027926.1 4788 UTR 3 XP_016883415.1

View Full Product Details