Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001284260.1 | Intron | NP_001271189.1 | |||
NM_001284261.1 | Intron | NP_001271190.1 | |||
NM_001284262.1 | Intron | NP_001271191.1 | |||
NM_198285.2 | Intron | NP_938026.2 | |||
XM_005249989.3 | Intron | XP_005250046.1 | |||
XM_005249990.4 | Intron | XP_005250047.1 | |||
XM_006715966.2 | Intron | XP_006716029.1 | |||
XM_011516144.2 | Intron | XP_011514446.1 | |||
XM_011516145.2 | Intron | XP_011514447.1 | |||
XM_011516146.2 | Intron | XP_011514448.1 | |||
XM_011516147.2 | Intron | XP_011514449.1 | |||
XM_011516148.1 | Intron | XP_011514450.1 | |||
XM_011516150.1 | Intron | XP_011514452.1 | |||
XM_011516151.2 | Intron | XP_011514453.1 | |||
XM_011516152.1 | Intron | XP_011514454.1 |