Product Details

SNP ID
rs798675
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:47610 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTAAAAAAAGATGCTACTCATG[A/G]CTGTCAAAAGTAGGTATTACTTAAT
Phenotype
MIM: 606007
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
POLR3K PubMed Links

Gene Details

Gene
POLR3K
Gene Name
RNA polymerase III subunit K
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016310.4 Intron NP_057394.3
Gene
SNRNP25
Gene Name
small nuclear ribonucleoprotein U11/U12 subunit 25
There are no transcripts associated with this gene.

View Full Product Details