Product Details

SNP ID
rs200553495
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:103277342 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGGCTTCCGCTCGCAGTCGCTGT[C/T]CCGCAGCAATGTGGCCTCCTCGGCC
Phenotype
MIM: 605338
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
INA PubMed Links

Gene Details

Gene
INA
Gene Name
internexin neuronal intermediate filament protein alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032727.3 180 Missense Mutation TCC,TTC S44F NP_116116.1

View Full Product Details