Product Details

SNP ID
rs200288509
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65573401 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTGGACATGTCCTTGTTGTGCCA[C/G]CTGTGGGGCCTGTACGAGTCAATCC
Phenotype
MIM: 616128 MIM: 606044
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EHBP1L1 PubMed Links

Gene Details

Gene
EHBP1L1
Gene Name
EH domain binding protein 1 like 1
There are no transcripts associated with this gene.

Gene
FAM89B
Gene Name
family with sequence similarity 89 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098784.1 651 Missense Mutation CCT,GCT P114A NP_001092254.1
NM_001098785.1 651 Missense Mutation CAC,CAG H110Q NP_001092255.1
NM_152832.2 651 Intron NP_690045.1
Gene
SSSCA1
Gene Name
Sjogren syndrome/scleroderma autoantigen 1
There are no transcripts associated with this gene.

Gene
SSSCA1-AS1
Gene Name
SSSCA1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details