Product Details

SNP ID
rs200043601
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64556184 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGACAATGGCTCTGCGGTTTCCA[C/G]AAACATGACCCCCAAGGCCCTTCTG
Phenotype
MIM: 607097
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SLC22A11 PubMed Links

Gene Details

Gene
SLC22A11
Gene Name
solute carrier family 22 member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307985.1 584 Missense Mutation ACA,AGA T62R NP_001294914.1
NM_018484.3 584 Missense Mutation ACA,AGA T62R NP_060954.1
XM_011545167.1 584 UTR 5 XP_011543469.1

View Full Product Details