Product Details

SNP ID
rs200593751
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64556135 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGAACTTCTCAGCCGCCATCCCA[A/G]GCCACCGATGCTGGACACACATGCT
Phenotype
MIM: 607097
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC22A11 PubMed Links

Gene Details

Gene
SLC22A11
Gene Name
solute carrier family 22 member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307985.1 535 Missense Mutation AGC,GGC S46G NP_001294914.1
NM_018484.3 535 Missense Mutation AGC,GGC S46G NP_060954.1
XM_011545167.1 535 UTR 5 XP_011543469.1

View Full Product Details