Product Details

SNP ID
rs201551635
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124424240 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGCGTAAGAGATGACGATGCTT[A/G]TGCTGGATAGCATGGTGATTACTCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR8B4 PubMed Links

Gene Details

Gene
OR8B4
Gene Name
olfactory receptor family 8 subfamily B member 4 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005196.1 632 Missense Mutation NP_001005196.1

View Full Product Details