Product Details

SNP ID
rs201311974
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60919763 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGCCATTCTGATGGTCCTAGTG[C/G]CCCTTATCCTCCTCCACTCAGCATT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TMEM109 PubMed Links

Gene Details

Gene
TMEM109
Gene Name
transmembrane protein 109
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024092.2 448 Missense Mutation CCC,GCC P24A NP_076997.1
Gene
TMEM132A
Gene Name
transmembrane protein 132A
There are no transcripts associated with this gene.

View Full Product Details