Product Details

SNP ID
rs201123323
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:113410919 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCAGCCAGCAGATGATGAACAC[A/G]CCTGGGGGAGAGGGCATGGTCAGGC
Phenotype
MIM: 126450
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DRD2 PubMed Links

Gene Details

Gene
DRD2
Gene Name
dopamine receptor D2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000795.3 1357 Silent Mutation GGC,GGT G380G NP_000786.1
NM_016574.3 1357 Silent Mutation GGC,GGT G351G NP_057658.2
XM_017017296.1 1357 Silent Mutation GGC,GGT G380G XP_016872785.1

View Full Product Details