Product Details

SNP ID
rs201081087
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57182207 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCGGCTATTCTCCGTGCCCCCAG[A/C]CCTGCCAATGGACACCCGAAACCTC
Phenotype
MIM: 615213
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LRRC55 PubMed Links

Gene Details

Gene
LRRC55
Gene Name
leucine rich repeat containing 55
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005210.2 461 Missense Mutation GAC,GCC D105A NP_001005210.1

View Full Product Details