Product Details

SNP ID
rs202035356
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52233560 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGACCCCTCCCTCCAGCGGGTGCG[C/T]CAGGAGGAGAGCGAGCAGATCAAGA
Phenotype
MIM: 148059
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRT7 PubMed Links

Gene Details

Gene
KRT7
Gene Name
keratin 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005556.3 344 Silent Mutation CGC,CGT R88R NP_005547.3
XM_011538325.1 344 Silent Mutation CGC,CGT R88R XP_011536627.1
XM_017019294.1 344 Intron XP_016874783.1
Gene
LINC00592
Gene Name
long intergenic non-protein coding RNA 592
There are no transcripts associated with this gene.

View Full Product Details