Product Details

SNP ID
rs200691357
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.13:38687904 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGAGGTTGTGACTCGGAACTTGC[C/T]TCTGGTCGTGGAAGAGCTGCTGGGG
Phenotype
MIM: 608945
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
FREM2 PubMed Links

Gene Details

Gene
FREM2
Gene Name
FRAS1 related extracellular matrix protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207361.5 869 Missense Mutation CCT,CTT P187L NP_997244.4
XM_017020554.1 869 Missense Mutation CCT,CTT P187L XP_016876043.1
Gene
LINC00437
Gene Name
long intergenic non-protein coding RNA 437
There are no transcripts associated with this gene.

View Full Product Details