Product Details

SNP ID
rs201313418
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:27548440 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGCTCATGCCCACGGTTGACAG[A/C]CCATTTACGGCCACAATCATGTCAC
Phenotype
MIM: 609733
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LNX2 PubMed Links

Gene Details

Gene
LNX2
Gene Name
ligand of numb-protein X 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153371.3 2136 Silent Mutation GGG,GGT G656G NP_699202.1
XM_011534995.2 2136 Silent Mutation GGG,GGT G533G XP_011533297.1
XM_017020434.1 2136 Silent Mutation GGG,GGT G656G XP_016875923.1

View Full Product Details