Product Details

SNP ID
hCV190008345
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24233581 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATACTGTGGGCACCTTTGAGATG[C/G]CCAAGGTTCTCTGTAAGGTAGGGCT
Phenotype
MIM: 610781 MIM: 603171 MIM: 604319
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
GMPR2 PubMed Links
Additional Information
For this assay, SNP(s) [rs2295316] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
GMPR2
Gene Name
guanosine monophosphate reductase 2
There are no transcripts associated with this gene.

Gene
NEDD8
Gene Name
neural precursor cell expressed, developmentally down-regulated 8
There are no transcripts associated with this gene.

Gene
NEDD8-MDP1
Gene Name
NEDD8-MDP1 readthrough
There are no transcripts associated with this gene.

Gene
TINF2
Gene Name
TERF1 interacting nuclear factor 2
There are no transcripts associated with this gene.

View Full Product Details