Product Details

SNP ID
rs200546417
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:113820952 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCTCGTCCAGGTCCAGCAGCCTC[T/C]GGTTGACCTCCAGTGTCATGCAGCC
Phenotype
MIM: 600441
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
GAS6 PubMed Links

Gene Details

Gene
GAS6
Gene Name
growth arrest specific 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000820.3 2132 Missense Mutation CAG,CGG Q650R NP_000811.1
Gene
GAS6-AS1
Gene Name
GAS6 antisense RNA 1
There are no transcripts associated with this gene.

Gene
TMEM255B
Gene Name
transmembrane protein 255B
There are no transcripts associated with this gene.

View Full Product Details