Product Details

SNP ID
rs200816634
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:91272778 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGAGACCGACCTGGGCTCTTGGC[C/T]GGAAGCCCCTCACTGCAGCCCTGCC
Phenotype
MIM: 611204 MIM: 601404
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC88C PubMed Links

Gene Details

Gene
CCDC88C
Gene Name
coiled-coil domain containing 88C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080414.3 3151 Silent Mutation CCA,CCG P1978P NP_001073883.2
XM_005267691.4 3151 Intron XP_005267748.1
XM_011536796.2 3151 Silent Mutation CCA,CCG P1942P XP_011535098.1
XM_017021335.1 3151 Intron XP_016876824.1
XM_017021336.1 3151 Silent Mutation CCA,CCG P1005P XP_016876825.1
XM_017021337.1 3151 Intron XP_016876826.1
Gene
GPR68
Gene Name
G protein-coupled receptor 68
There are no transcripts associated with this gene.

View Full Product Details