Product Details

SNP ID
rs200253000
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43874190 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCCCCAAGGGCCTCCACCTCTG[C/T]AGCAGTTGGGGTGCTGGCTTCAGAT
Phenotype
MIM: 616309
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
FRMD5 PubMed Links

Gene Details

Gene
FRMD5
Gene Name
FERM domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286490.1 1648 Missense Mutation ACA,GCA T376A NP_001273419.1
NM_001286491.1 1648 Missense Mutation ACA,GCA T236A NP_001273420.1
NM_001322949.1 1648 Missense Mutation ACA,GCA T470A NP_001309878.1
NM_001322950.1 1648 Missense Mutation ACA,GCA T470A NP_001309879.1
NM_001322951.1 1648 Missense Mutation ACA,GCA T435A NP_001309880.1
NM_032892.4 1648 Missense Mutation ACA,GCA T470A NP_116281.2
XM_005254730.3 1648 Missense Mutation ACA,GCA T470A XP_005254787.1
Gene
PIN4P1
Gene Name
peptidylprolyl cis/trans isomerase, NIMA-interacting 4 pseudogene 1
There are no transcripts associated with this gene.

Gene
WDR76
Gene Name
WD repeat domain 76
There are no transcripts associated with this gene.

View Full Product Details