Product Details

SNP ID
rs200120066
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:57976518 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTCCTTTATTCTGCTGCTGTCCA[C/T]AATAAGGGCTCGTCCACCTCCCTTC
Phenotype
MIM: 600724 MIM: 610264
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNGB1 PubMed Links

Gene Details

Gene
CNGB1
Gene Name
cyclic nucleotide gated channel beta 1
There are no transcripts associated with this gene.

Gene
TEPP
Gene Name
testis, prostate and placenta expressed
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199046.2 84 Missense Mutation ACA,ATA T16I NP_950247.2
NM_199456.2 84 Missense Mutation ACA,ATA T16I NP_955535.2
XM_017023218.1 84 Intron XP_016878707.1

View Full Product Details