Product Details

SNP ID
rs201665846
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29744299 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTCAGAACCCCACACTGGTCCGG[A/C]CTTCACGCTTCCAGAAAGCTGAGAT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C16orf54 PubMed Links

Gene Details

Gene
C16orf54
Gene Name
chromosome 16 open reading frame 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175900.3 762 Missense Mutation NP_787096.2

View Full Product Details