Product Details
- SNP ID
-
rs201908654
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:11273390 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGCAGCAGCAGCTTGCCCTGCACC[A/G]GCAGCTCTTCTTTCTCCTCCTCTTC
- Phenotype
-
MIM: 182880
MIM: 182890
MIM: 190232
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PRM1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs35598356] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PRM1
- Gene Name
- protamine 1
There are no transcripts associated with this gene.
- Gene
- PRM2
- Gene Name
- protamine 2
There are no transcripts associated with this gene.
- Gene
- PRM3
- Gene Name
- protamine 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021247.2 |
206 |
Missense Mutation |
CCG,CTG |
P69L |
NP_067070.2 |
- Gene
- TNP2
- Gene Name
- transition protein 2 (during histone to protamine replacement)
There are no transcripts associated with this gene.
View Full Product Details