Product Details
- SNP ID
-
rs200313002
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:7688852 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTCAGCTGTGTCCCAGGAGCTACG[C/G]GAGGGGGACCCAGTTTCTCTCTCCA
- Phenotype
-
MIM: 191170
MIM: 612661
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TP53
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2287499] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TP53
- Gene Name
- tumor protein p53
There are no transcripts associated with this gene.
- Gene
- WRAP53
- Gene Name
- WD repeat containing antisense to TP53
View Full Product Details