Product Details

SNP ID
rs199502613
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40184645 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGCCTCCGAGAGGACACTCTGA[A/G]GCTGCACCAGCTGGTGGAGACGGTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR6867 PubMed Links

Gene Details

Gene
MIR6867
Gene Name
microRNA 6867
There are no transcripts associated with this gene.

Gene
RAPGEFL1
Gene Name
Rap guanine nucleotide exchange factor like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303533.1 28 Missense Mutation AAG,AGG K116R NP_001290462.1
NM_001303534.1 28 Missense Mutation AAG,AGG K110R NP_001290463.1
NM_016339.4 28 Missense Mutation AAG,AGG K61R NP_057423.1
XM_006721932.3 28 Missense Mutation AAG,AGG K267R XP_006721995.1
XM_006721935.3 28 Missense Mutation AAG,AGG K98R XP_006721998.1
XM_006721937.3 28 Missense Mutation AAG,AGG K61R XP_006722000.1
XM_006721938.3 28 UTR 5 XP_006722001.1

View Full Product Details