Product Details
- SNP ID
-
rs199651076
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:41583597 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCAATCTCCAGGTTCTGCATGGTG[C/T]GCCGGAGCTCCGAGATCTCGCTCTT
- Phenotype
-
MIM: 148066
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KRT14
PubMed Links
Gene Details
- Gene
- KRT14
- Gene Name
- keratin 14
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000526.4 |
|
Intron |
|
|
NP_000517.2 |
View Full Product Details