Product Details

SNP ID
rs200407214
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81245851 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCCCCCCGGGACAGCTCGTTTG[A/G]GCCCCAGCTCTGGCTCTGGCCTCTG
Phenotype
MIM: 616525
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C17orf89 PubMed Links

Gene Details

Gene
C17orf89
Gene Name
chromosome 17 open reading frame 89
There are no transcripts associated with this gene.

Gene
SLC38A10
Gene Name
solute carrier family 38 member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037984.2 2877 Missense Mutation CCC,CTC P1022L NP_001033073.1
NM_138570.3 2877 Intron NP_612637.1
XM_005257019.1 2877 Missense Mutation CCC,CTC P1030L XP_005257076.1
XM_011524288.1 2877 Missense Mutation CCC,CTC P1037L XP_011522590.1
XM_011524289.1 2877 Missense Mutation CCC,CTC P1029L XP_011522591.1
XM_011524290.1 2877 Missense Mutation CCC,CTC P948L XP_011522592.1
Gene
TEPSIN
Gene Name
TEPSIN, adaptor related protein complex 4 accessory protein
There are no transcripts associated with this gene.

View Full Product Details