Product Details

SNP ID
rs199728576
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:31937442 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTCCGCGGCGGCAGCGGCGGGG[A/G]CTGCGGTGTTACCGGTGAAGAAGCC
Phenotype
MIM: 606245
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SUZ12 PubMed Links

Gene Details

Gene
SUZ12
Gene Name
SUZ12 polycomb repressive complex 2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321207.1 433 Missense Mutation ACT,GCT T66A NP_001308136.1
NM_015355.3 433 Missense Mutation ACT,GCT T66A NP_056170.2
XM_006721794.3 433 Intron XP_006721857.1
XM_017024409.1 433 UTR 5 XP_016879898.1

View Full Product Details