Product Details

SNP ID
rs201024518
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:35352532 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAACACAATGTGATCACCCAACA[C/T]ATCACATGCATCACTGAGCTGCACC
Phenotype
MIM: 614953
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLFN11 PubMed Links

Gene Details

Gene
SLFN11
Gene Name
schlafen family member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001104587.1 2865 Intron NP_001098057.1
NM_001104588.1 2865 Intron NP_001098058.1
NM_001104589.1 2865 Intron NP_001098059.1
NM_001104590.1 2865 Intron NP_001098060.1
NM_152270.3 2865 Intron NP_689483.3
XM_005258068.3 2865 Missense Mutation ATG,GTG M844V XP_005258125.1
XM_006722171.3 2865 Missense Mutation ATG,GTG M844V XP_006722234.1
XM_011525465.2 2865 Missense Mutation ATG,GTG M844V XP_011523767.1
XM_017025299.1 2865 Missense Mutation ATG,GTG M844V XP_016880788.1

View Full Product Details