Product Details

SNP ID
rs200348981
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:5786484 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACTTACCGGGCAATCATGATCCC[A/G]GTGACACCAGTCTGCATGGCGCGGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CATSPERD PubMed Links

Gene Details

Gene
CATSPERD
Gene Name
cation channel sperm associated auxiliary subunit delta
There are no transcripts associated with this gene.

Gene
DUS3L
Gene Name
dihydrouridine synthase 3 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161619.1 1642 Silent Mutation ACC,ACT T273T NP_001155091.1
NM_020175.2 1642 Silent Mutation ACC,ACT T515T NP_064560.2
XM_017027020.1 1642 Silent Mutation ACC,ACT T501T XP_016882509.1
Gene
PRR22
Gene Name
proline rich 22
There are no transcripts associated with this gene.

View Full Product Details