Product Details

SNP ID
rs199570925
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:6426444 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACACCACATAGCTGATGCCACCT[A/G]CTGGTAAGACCTTCAGTAGCGTGGG
Phenotype
MIM: 603445 MIM: 608746 MIM: 610822
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KHSRP PubMed Links

Gene Details

Gene
KHSRP
Gene Name
KH-type splicing regulatory protein
There are no transcripts associated with this gene.

Gene
MIR3940
Gene Name
microRNA 3940
There are no transcripts associated with this gene.

Gene
SLC25A23
Gene Name
solute carrier family 25 member 23
There are no transcripts associated with this gene.

Gene
SLC25A41
Gene Name
solute carrier family 25 member 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321298.1 938 Missense Mutation GCA,GTA A215V NP_001308227.1
NM_173637.3 938 Missense Mutation GCA,GTA A353V NP_775908.2
XM_011527926.1 938 Missense Mutation GCA,GTA A289V XP_011526228.1

View Full Product Details