Product Details

SNP ID
rs200270910
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2757762 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGTTCCCAAGGGGTTGTTGCCA[A/C]CCGAAATCATGCCGGACATGCTGCA
Phenotype
MIM: 603419
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SGTA PubMed Links

Gene Details

Gene
SGTA
Gene Name
small glutamine rich tetratricopeptide repeat containing alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003021.3 959 Missense Mutation NP_003012.1
XM_011528178.2 959 Missense Mutation XP_011526480.1

View Full Product Details