Product Details

SNP ID
rs199580884
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11723215 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATTAAGAGACGAATGACCCAAGA[C/T]GACTTCTCCACACTCACCACTGTCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF823 PubMed Links

Gene Details

Gene
ZNF823
Gene Name
zinc finger protein 823
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080493.3 382 Missense Mutation ATC,GTC I107V NP_001073962.1
NM_001297610.1 382 UTR 5 NP_001284539.1
NM_017507.1 382 Missense Mutation ATC,GTC I63V NP_059977.1
XM_017026941.1 382 UTR 5 XP_016882430.1
XM_017026942.1 382 UTR 5 XP_016882431.1

View Full Product Details