Product Details

SNP ID
rs201799125
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2757376 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGTCGTCGTTGCTGGCGCTGGG[C/T]GTCCGACTCCGGATCTGGCTCCTGA
Phenotype
MIM: 603419
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SGTA PubMed Links

Gene Details

Gene
SGTA
Gene Name
small glutamine rich tetratricopeptide repeat containing alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003021.3 1110 Silent Mutation ACA,ACG T303T NP_003012.1
XM_011528178.2 1110 Silent Mutation ACA,ACG T303T XP_011526480.1

View Full Product Details