Product Details
- SNP ID
-
rs202219548
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:19219038 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCTGTCTCTTTACCCTGCAGCCAC[A/G]GCCAAGCGCAGCCCGAGATGCCCCT
- Phenotype
-
MIM: 600826
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NCAN
PubMed Links
Gene Details
- Gene
- NCAN
- Gene Name
- neurocan
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004386.2 |
296 |
Missense Mutation |
CAG,CGG |
Q66R |
NP_004377.2 |
View Full Product Details