Product Details

SNP ID
rs201767859
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11343095 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCGGGCGTTGACAGTGGCCAGC[A/G]TAAGGCTCAGGAACAGCAGGTAAAG
Phenotype
MIM: 604350 MIM: 613771
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC159 PubMed Links

Gene Details

Gene
CCDC159
Gene Name
coiled-coil domain containing 159
There are no transcripts associated with this gene.

Gene
RAB3D
Gene Name
RAB3D, member RAS oncogene family
There are no transcripts associated with this gene.

Gene
TMEM205
Gene Name
transmembrane protein 205
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145416.1 288 Missense Mutation ACG,ATG T97M NP_001138888.1
NM_001321112.1 288 Missense Mutation ACG,ATG T97M NP_001308041.1
NM_001321113.1 288 Missense Mutation ACG,ATG T97M NP_001308042.1
NM_001321114.1 288 Missense Mutation ACG,ATG T97M NP_001308043.1
NM_033408.3 288 Missense Mutation ACG,ATG T97M NP_212133.1
NM_198536.2 288 Missense Mutation ACG,ATG T97M NP_940938.1
XM_005259899.2 288 Missense Mutation ACG,ATG T97M XP_005259956.1
XM_017026769.1 288 Silent Mutation TAC,TAT Y90Y XP_016882258.1

View Full Product Details