Product Details

SNP ID
rs200093391
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:70221651 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGCAGCGCCATGAGCAACACTAC[A/C]GTCGTCCCCAGCACTGCAGGTCCGG
Phenotype
MIM: 602010
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SRSF11 PubMed Links

Gene Details

Gene
SRSF11
Gene Name
serine and arginine rich splicing factor 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190987.1 136 Silent Mutation ACA,ACC T5T NP_001177916.1
NM_004768.3 136 Silent Mutation ACA,ACC T5T NP_004759.1
XM_005271338.3 136 Silent Mutation ACA,ACC T5T XP_005271395.1
XM_005271339.3 136 Silent Mutation ACA,ACC T5T XP_005271396.1
XM_011542427.2 136 Intron XP_011540729.1
XM_011542428.2 136 Intron XP_011540730.1
XM_011542431.2 136 UTR 5 XP_011540733.1
XM_017002811.1 136 Silent Mutation ACA,ACC T5T XP_016858300.1
XM_017002812.1 136 Silent Mutation ACA,ACC T5T XP_016858301.1
XM_017002813.1 136 Silent Mutation ACA,ACC T5T XP_016858302.1
XM_017002814.1 136 Intron XP_016858303.1
XM_017002815.1 136 Intron XP_016858304.1
XM_017002816.1 136 UTR 5 XP_016858305.1
XM_017002817.1 136 Intron XP_016858306.1
XM_017002818.1 136 Intron XP_016858307.1
XM_017002819.1 136 Intron XP_016858308.1
XM_017002820.1 136 Intron XP_016858309.1
XM_017002821.1 136 Intron XP_016858310.1
XM_017002822.1 136 Intron XP_016858311.1
XM_017002823.1 136 Intron XP_016858312.1
XM_017002824.1 136 UTR 5 XP_016858313.1
XM_017002825.1 136 Intron XP_016858314.1
XM_017002826.1 136 Intron XP_016858315.1
XM_017002827.1 136 Intron XP_016858316.1
XM_017002828.1 136 UTR 5 XP_016858317.1
XM_017002829.1 136 UTR 5 XP_016858318.1
XM_017002830.1 136 UTR 5 XP_016858319.1
XM_017002831.1 136 Intron XP_016858320.1
XM_017002832.1 136 Intron XP_016858321.1
XM_017002833.1 136 Intron XP_016858322.1
XM_017002834.1 136 Intron XP_016858323.1

View Full Product Details