Product Details

SNP ID
rs200283561
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:113980605 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGGGAAGAAGGAATGAGAAGTAC[A/G]ATATGGTGACAGGTAAATAATCTGA
Phenotype
MIM: 608003 MIM: 610088
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HIPK1 PubMed Links

Gene Details

Gene
HIPK1
Gene Name
homeodomain interacting protein kinase 1
There are no transcripts associated with this gene.

Gene
OLFML3
Gene Name
olfactomedin like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286352.1 666 Missense Mutation AAT,GAT N110D NP_001273281.1
NM_001286353.1 666 Missense Mutation AAT,GAT N69D NP_001273282.1
NM_020190.3 666 Missense Mutation AAT,GAT N130D NP_064575.1
XM_017001848.1 666 Missense Mutation AAT,GAT N110D XP_016857337.1

View Full Product Details