Product Details
- SNP ID
-
rs200179940
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
5
- Location
-
Chr.1:89009055 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATGTTCTTGATAACTCTTCTCTTT[C/G]TCTTCCATCATCTGCTGATACTTTA
- Phenotype
-
MIM: 600413
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GBP3
PubMed Links
Gene Details
- Gene
- GBP3
- Gene Name
- guanylate binding protein 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001319179.1 |
1689 |
Missense Mutation |
GAC,GAG |
D438E |
NP_001306108.1 |
NM_001319180.1 |
1689 |
Missense Mutation |
GAC,GAG |
D383E |
NP_001306109.1 |
NM_001319181.1 |
1689 |
Missense Mutation |
GAC,GAG |
D490E |
NP_001306110.1 |
NM_018284.2 |
1689 |
Missense Mutation |
GAC,GAG |
D517E |
NP_060754.2 |
XM_005270743.2 |
1689 |
Missense Mutation |
GAC,GAG |
D438E |
XP_005270800.1 |
XM_006710574.2 |
1689 |
Missense Mutation |
GAC,GAG |
D490E |
XP_006710637.1 |
XM_011541233.1 |
1689 |
Missense Mutation |
GAC,GAG |
D438E |
XP_011539535.1 |
XM_011541234.1 |
1689 |
Missense Mutation |
GAC,GAG |
D436E |
XP_011539536.1 |
XM_017001013.1 |
1689 |
Missense Mutation |
GAC,GAG |
D383E |
XP_016856502.1 |
XM_017001014.1 |
1689 |
Missense Mutation |
GAC,GAG |
D302E |
XP_016856503.1 |
View Full Product Details