Product Details

SNP ID
rs201971593
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:70221837 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGACGAACTGCGCCTCTTCCCGCC[A/G]GAGTGAGTATCGTCCACCATCACTG
Phenotype
MIM: 602010
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SRSF11 PubMed Links

Gene Details

Gene
SRSF11
Gene Name
serine and arginine rich splicing factor 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190987.1 322 Silent Mutation CCA,CCG P67P NP_001177916.1
NM_004768.3 322 Silent Mutation CCA,CCG P67P NP_004759.1
XM_005271338.3 322 Silent Mutation CCA,CCG P67P XP_005271395.1
XM_005271339.3 322 Silent Mutation CCA,CCG P67P XP_005271396.1
XM_011542427.2 322 Intron XP_011540729.1
XM_011542428.2 322 Intron XP_011540730.1
XM_011542431.2 322 UTR 5 XP_011540733.1
XM_017002811.1 322 Silent Mutation CCA,CCG P67P XP_016858300.1
XM_017002812.1 322 Silent Mutation CCA,CCG P67P XP_016858301.1
XM_017002813.1 322 Silent Mutation CCA,CCG P67P XP_016858302.1
XM_017002814.1 322 Intron XP_016858303.1
XM_017002815.1 322 Intron XP_016858304.1
XM_017002816.1 322 UTR 5 XP_016858305.1
XM_017002817.1 322 Intron XP_016858306.1
XM_017002818.1 322 Intron XP_016858307.1
XM_017002819.1 322 Intron XP_016858308.1
XM_017002820.1 322 Intron XP_016858309.1
XM_017002821.1 322 Intron XP_016858310.1
XM_017002822.1 322 Intron XP_016858311.1
XM_017002823.1 322 Intron XP_016858312.1
XM_017002824.1 322 UTR 5 XP_016858313.1
XM_017002825.1 322 Intron XP_016858314.1
XM_017002826.1 322 Intron XP_016858315.1
XM_017002827.1 322 Intron XP_016858316.1
XM_017002828.1 322 UTR 5 XP_016858317.1
XM_017002829.1 322 UTR 5 XP_016858318.1
XM_017002830.1 322 UTR 5 XP_016858319.1
XM_017002831.1 322 Intron XP_016858320.1
XM_017002832.1 322 Intron XP_016858321.1
XM_017002833.1 322 Intron XP_016858322.1
XM_017002834.1 322 Intron XP_016858323.1

View Full Product Details