Product Details

SNP ID
rs202016325
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:112701704 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCATGTCCCGGCCTTCCTCAGACC[G/T]AACGGGCTCCTGAGAAGACATAAGA
Phenotype
MIM: 610742 MIM: 609957 MIM: 165380
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MOV10 PubMed Links

Gene Details

Gene
MOV10
Gene Name
Mov10 RISC complex RNA helicase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130079.2 4659 Intron NP_001123551.1
NM_001286072.1 4659 Intron NP_001273001.1
NM_001321324.1 4659 Intron NP_001308253.1
NM_020963.4 4659 Intron NP_066014.1
XM_005270869.4 4659 UTR 3 XP_005270926.1
XM_005270870.4 4659 UTR 3 XP_005270927.1
XM_017001318.1 4659 UTR 3 XP_016856807.1
XM_017001319.1 4659 Intron XP_016856808.1
Gene
PPM1J
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1J
There are no transcripts associated with this gene.

Gene
RHOC
Gene Name
ras homolog family member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042678.1 4659 Silent Mutation NP_001036143.1
NM_001042679.1 4659 Silent Mutation NP_001036144.1
NM_175744.4 4659 Silent Mutation NP_786886.1

View Full Product Details