Product Details

SNP ID
rs202055717
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:27967018 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCTGGGCCTGGCTCTGCGGCTT[T/G]TGTACTACCACTGGCTGCACCCTAG
Phenotype
MIM: 601655
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
EYA3 PubMed Links

Gene Details

Gene
EYA3
Gene Name
EYA transcriptional coactivator and phosphatase 3
There are no transcripts associated with this gene.

Gene
SMPDL3B
Gene Name
sphingomyelin phosphodiesterase acid like 3B
There are no transcripts associated with this gene.

Gene
XKR8
Gene Name
XK related 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018053.3 1614 Missense Mutation GTG,TTG V336L NP_060523.2
XM_011541679.2 1614 Missense Mutation GTG,TTG V390L XP_011539981.1
XM_011541680.2 1614 Missense Mutation GTG,TTG V354L XP_011539982.1

View Full Product Details