Product Details

SNP ID
rs201539344
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:25418117 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGATGTCCTAGGTATGACCGCTT[A/G]CTTCGGATCAGAGCACTCAGATGGG
Phenotype
MIM: 610608
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GINS1 PubMed Links

Gene Details

Gene
GINS1
Gene Name
GINS complex subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021067.4 347 Silent Mutation TTA,TTG L84L NP_066545.3
XM_017028162.1 347 Silent Mutation TTA,TTG L49L XP_016883651.1

View Full Product Details